Assay Details
| Gene Symbol: CKMT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695305.3 | XP_005695362.1 | 2 - 3 | 418 | 71 | |
| XM_018066415.1 | XP_017921904.1 | 2 - 3 | 724 | 71 | ||
| XM_018066418.1 | XP_017921907.1 | 2 - 3 | 408 | 71 | ||
| XM_018066419.1 | XP_017921908.1 | 2 - 3 | 425 | 71 | ||
| GenBank mRNA | KA309735.1 | - | 2 - 3 | 511 | 71 | |
More Information
Set Membership: |
|

CKMT1A
Genomic Map