Assay Details
| Gene Symbol: ECHS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041274.1 | XP_017896763.1 | 6 - 7 | 806 | 62 | |
| GenBank mRNA | JO560234.2 | - | 6 - 7 | 1025 | 62 | |
| JO578203.1 | - | 6 - 7 | 845 | 62 | ||
| KA326402.1 | - | 6 - 7 | 775 | 62 | ||
More Information
Set Membership: |
|

ECHS1
Genomic Map