Assay Details
| Gene Symbol: PPP1R14B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699968.3 | XP_005700025.3 | 1 - 2 | 513 | 91 | |
| GenBank mRNA | JO510384.2 | - | 3 - 4 | 446 | 91 | |
More Information
Set Membership: |
|

PPP1R14B
Genomic Map