Assay Details
| Gene Symbol: LOC102183306 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049273.1 | XP_017904762.1 | 2 - 3 | 296 | 71 | |
| GenBank mRNA | JO404007.2 | - | 2 - 3 | 283 | 71 | |
| JO579992.1 | - | 2 - 3 | 353 | 71 | ||
More Information
Set Membership: |
|

LOC102183306
Genomic Map