Assay Details
| Gene Symbol: LATS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053335.1 | XP_017908824.1 | 3 - 4 | 998 | 76 | |
| XM_018053336.1 | XP_017908825.1 | 3 - 4 | 998 | 76 | ||
More Information
Set Membership: |
|

LATS1
Genomic Map