Assay Details
| Gene Symbol: CCDC187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056087.1 | XP_017911576.1 | 2 - 3 | 594 | 93 | |
| XM_018056088.1 | XP_017911577.1 | 2 - 3 | 594 | 93 | ||
| XM_018056089.1 | XP_017911578.1 | 2 - 3 | 594 | 93 | ||
| XM_018056091.1 | XP_017911580.1 | 2 - 3 | 594 | 93 | ||
| XM_018056092.1 | XP_017911581.1 | 2 - 3 | 594 | 93 | ||
More Information
Set Membership: |
|

CCDC187
Genomic Map