Assay Details
| Gene Symbol: CCDC187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056087.1 | XP_017911576.1 | 5 - 6 | 1946 | 79 | |
| XM_018056088.1 | XP_017911577.1 | 5 - 6 | 1946 | 79 | ||
| XM_018056089.1 | XP_017911578.1 | 5 - 6 | 1946 | 79 | ||
| XM_018056090.1 | XP_017911579.1 | 4 - 5 | 1850 | 79 | ||
| XM_018056091.1 | XP_017911580.1 | 5 - 6 | 1946 | 79 | ||
| XM_018056092.1 | XP_017911581.1 | 5 - 6 | 1946 | 79 | ||
More Information
Set Membership: |
|

CCDC187
Genomic Map