Assay Details
| Gene Symbol: RPL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013969042.2 | XP_013824496.2 | 3 - 4 | 424 | 65 | |
| GenBank mRNA | KA340345.1 | - | 3 - 4 | 535 | 65 | |
More Information
Set Membership: |
|

RPL8
Genomic Map