Assay Details
| Gene Symbol: NFATC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039658.1 | XP_017895147.1 | 7 - 8 | 2272 | 70 | |
| XM_018039659.1 | XP_017895148.1 | 7 - 8 | 2219 | 70 | ||
| XM_018039660.1 | XP_017895149.1 | 7 - 8 | 2271 | 70 | ||
| XM_018039661.1 | XP_017895150.1 | 7 - 8 | 2219 | 70 | ||
| XM_018039662.1 | XP_017895151.1 | 7 - 8 | 2270 | 70 | ||
| XM_018039663.1 | XP_017895152.1 | 7 - 8 | 2269 | 70 | ||
| GenBank mRNA | JO599218.2 | - | 3 - 4 | 2631 | 70 | |
| JO599219.2 | - | 3 - 4 | 2631 | 70 | ||
More Information
Set Membership: |
|

NFATC1
Genomic Map