Assay Details
| Gene Symbol: PDPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040359.1 | XP_017895848.1 | 10 - 11 | 1259 | 74 | |
| XM_018040360.1 | XP_017895849.1 | 9 - 10 | 1178 | 74 | ||
| GenBank mRNA | JO426018.1 | - | 10 - 11 | 1322 | 74 | |
More Information
Set Membership: |
|

PDPK1
Genomic Map