Assay Details
| Gene Symbol: VWA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041456.1 | XP_017896945.1 | 10 - 11 | 1513 | 58 | |
| XM_018041457.1 | XP_017896946.1 | 10 - 11 | 1513 | 58 | ||
More Information
Set Membership: |
|

VWA2
Genomic Map