Assay Details
| Gene Symbol: CALY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041714.1 | XP_017897203.1 | 2 - 3 | 250 | 67 | |
| XM_018041715.1 | XP_017897204.1 | 2 - 3 | 250 | 67 | ||
| XM_018041716.1 | XP_017897205.1 | 2 - 3 | 409 | 67 | ||
| GenBank mRNA | JO634921.2 | - | 2 - 3 | 562 | 67 | |
| JO634929.2 | - | 2 - 3 | 562 | 67 | ||
More Information
Set Membership: |
|

CALY
Genomic Map