Assay Details
Gene Symbol: RETREG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059226.1 | XP_017914715.1 | 7 - 8 | 991 | 68 | |
XM_018059231.1 | XP_017914720.1 | 7 - 8 | 907 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
