Assay Details
| Gene Symbol: ETV3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046332.1 | XP_017901821.1 | 3 - 4 | 571 | 83 | |
| XM_018046333.1 | XP_017901822.1 | 3 - 4 | 455 | 83 | ||
| XM_018046334.1 | XP_017901823.1 | 3 - 4 | 458 | 83 | ||
More Information
Set Membership: |
|

ETV3
Genomic Map