Assay Details
| Gene Symbol: C3H1orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677439.3 | XP_005677496.2 | 3 - 4 | 663 | 82 | |
| XM_018046187.1 | XP_017901676.1 | 2 - 3 | 561 | 82 | ||
| XM_018046188.1 | XP_017901677.1 | 3 - 4 | 663 | 82 | ||
| XM_018046189.1 | XP_017901678.1 | 2 - 3 | 561 | 82 | ||
| GenBank mRNA | JO373438.2 | - | 1213 | 82 | ||
| JO499345.2 | - | 4 - 5 | 1222 | 82 | ||
| Gene Symbol: LOC102177858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040521.1 | XP_017896010.1 | 1 | 462 | 82 | |
| XM_018040522.1 | XP_017896011.1 | 2 | 360 | 82 | ||
More Information
Set Membership: |
|

C3H1orf43
Genomic Map