Assay Details
| Gene Symbol: FGGY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678344.2 | XP_005678401.2 | 4 - 5 | 576 | 61 | |
| XM_013962793.2 | XP_013818247.2 | 4 - 5 | 579 | 61 | ||
| XM_013962794.2 | XP_013818248.2 | 4 - 5 | 569 | 61 | ||
| XM_018044764.1 | XP_017900253.1 | 4 - 5 | 579 | 61 | ||
| GenBank mRNA | JO553351.2 | - | 4 - 5 | 557 | 61 | |
More Information
Set Membership: |
|

FGGY
Genomic Map