Assay Details
| Gene Symbol: CPM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047966.1 | XP_017903455.1 | 8 - 9 | 1167 | 66 | |
| XM_018047967.1 | XP_017903456.1 | 7 - 8 | 1048 | 66 | ||
More Information
Set Membership: |
|

CPM
Genomic Map