Assay Details
| Gene Symbol: IKBIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680481.3 | XP_005680538.2 | 1 - 2 | 289 | 81 | |
| XM_018048136.1 | XP_017903625.1 | 1 - 2 | 404 | 81 | ||
| GenBank mRNA | JO583786.1 | - | 2 - 3 | 1033 | 81 | |
More Information
Set Membership: |
|

IKBIP
Genomic Map