Assay Details
| Gene Symbol: CENPM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681122.3 | XP_005681179.1 | 5 - 6 | 478 | 69 | |
| GenBank mRNA | JO598477.2 | - | 1 - 2 | 167 | 69 | |
More Information
Set Membership: |
|

CENPM
Genomic Map