Assay Details
| Gene Symbol: SRFBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682717.3 | XP_005682774.1 | 2 - 3 | 215 | 72 | |
| GenBank mRNA | JO596357.2 | - | 6 - 7 | 1173 | 72 | |
More Information
Set Membership: |
|

SRFBP1
Genomic Map