Assay Details
| Gene Symbol: XRCC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683394.3 | XP_005683451.1 | 2 - 3 | 311 | 63 | |
| GenBank mRNA | KA314498.1 | - | 6 - 7 | 1197 | 63 | |
More Information
Set Membership: |
|

XRCC4
Genomic Map