Assay Details
| Gene Symbol: SEPHS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687952.3 | XP_005688009.1 | 7 - 8 | 1029 | 60 | |
| XM_018057121.1 | XP_017912610.1 | 7 - 8 | 1147 | 60 | ||
More Information
Set Membership: |
|

SEPHS1
Genomic Map