Assay Details
| Gene Symbol: TGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688984.2 | XP_005689041.2 | 5 - 6 | 1440 | 65 | |
| XM_005688986.3 | XP_005689043.2 | 5 - 6 | 1461 | 65 | ||
| XM_013969193.2 | XP_013824647.2 | 5 - 6 | 1538 | 65 | ||
| XM_018058503.1 | XP_017913992.1 | 5 - 6 | 1377 | 65 | ||
More Information
Set Membership: |
|

TGS1
Genomic Map