Assay Details
| Gene Symbol: PRR5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013969814.2 | XP_013825268.1 | 6 - 7 | 1958 | 62 | |
| XM_013969817.2 | XP_013825271.1 | 4 - 5 | 1490 | 62 | ||
| XM_018059229.1 | XP_017914718.1 | 6 - 7 | 763 | 62 | ||
| XM_018059230.1 | XP_017914719.1 | 6 - 7 | 617 | 62 | ||
| XM_018059232.1 | XP_017914721.1 | 6 - 7 | 767 | 62 | ||
| GenBank mRNA | JO428789.2 | - | 6 - 7 | 819 | 62 | |
More Information
Set Membership: |
|

PRR5L
Genomic Map