Assay Details
| Gene Symbol: ARHGAP35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062659.1 | XP_017918148.1 | 3 - 4 | 4205 | 59 | |
| GenBank mRNA | JT678970.1 | - | 6 - 7 | 5639 | 59 | |
More Information
Set Membership: |
|

ARHGAP35
Genomic Map