Assay Details
| Gene Symbol: LOC102170645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038609.1 | XP_017894098.1 | 3 - 4 | 569 | 74 | |
| GenBank mRNA | JO436047.2 | - | 5 - 6 | 860 | 74 | |
More Information
Set Membership: |
|

LOC102170645
Genomic Map