Assay Details
| Gene Symbol: LOC102170645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038609.1 | XP_017894098.1 | 4 - 5 | 713 | 100 | |
| GenBank mRNA | JO436047.2 | - | 4 - 5 | 716 | 100 | |
More Information
Set Membership: |
|

LOC102170645
Genomic Map