Assay Details
| Gene Symbol: ZNRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696643.3 | XP_005696700.1 | 4 - 5 | 522 | 108 | |
| GenBank mRNA | JO578721.1 | - | 1 - 2 | 282 | 108 | |
More Information
Set Membership: |
|

ZNRD1
Genomic Map