Assay Details
| Gene Symbol: RBM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700785.3 | XP_005700842.1 | 2 - 3 | 245 | 72 | |
| XM_005700786.3 | XP_005700843.1 | 2 - 3 | 246 | 72 | ||
| XM_005700788.3 | XP_005700845.1 | 2 - 3 | 246 | 72 | ||
| XM_005700789.3 | XP_005700846.1 | 2 - 3 | 246 | 72 | ||
| XM_013976462.2 | XP_013831916.1 | 2 - 3 | 273 | 72 | ||
| GenBank mRNA | JO642884.2 | - | 5 - 6 | 528 | 72 | |
| JO642888.2 | - | 5 - 6 | 996 | 72 | ||
| KF933377.1 | - | 1 - 2 | 104 | 72 | ||
More Information
Set Membership: |
|

RBM3
Genomic Map