Assay Details
| Gene Symbol: BCL2L13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048707.1 | XP_017904196.1 | 3 - 4 | 387 | 69 | |
| XM_018048709.1 | XP_017904198.1 | 3 - 4 | 387 | 69 | ||
| XM_018048710.1 | XP_017904199.1 | 3 - 4 | 387 | 69 | ||
| XM_018048711.1 | XP_017904200.1 | 3 - 4 | 387 | 69 | ||
| GenBank mRNA | JO600244.2 | - | 3 - 4 | 408 | 69 | |
| JT689762.1 | - | 5 - 6 | 465 | 69 | ||
More Information
Set Membership: |
|

BCL2L13
Genomic Map