Assay Details
| Gene Symbol: ZNF367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052481.1 | XP_017907970.1 | 4 - 5 | 1252 | 67 | |
| XM_018052482.1 | XP_017907971.1 | 4 - 5 | 1104 | 67 | ||
More Information
Set Membership: |
|

ZNF367
Genomic Map