Assay Details
| Gene Symbol: LOC102173858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013968286.2 | XP_013823740.2 | 1 | 1431 | 203 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102173858| Gene Symbol: LOC102173858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013968286.2 | XP_013823740.2 | 1 | 1431 | 203 | |
Set Membership: |
|