Assay Details
| Gene Symbol: JPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058394.1 | XP_017913883.1 | 2 - 3 | 1408 | 70 | |
| XM_018058395.1 | XP_017913884.1 | 2 - 3 | 1408 | 70 | ||
| XM_018058396.1 | XP_017913885.1 | 2 - 3 | 1408 | 70 | ||
| XM_018058397.1 | XP_017913886.1 | 2 - 3 | 1408 | 70 | ||
| GenBank mRNA | JO630365.2 | - | 2177 | 70 | ||
More Information
Set Membership: |
|

JPH1
Genomic Map