Assay Details
| Gene Symbol: NRG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042553.1 | XP_017898042.1 | 5 - 6 | 1170 | 72 | |
| XM_018042554.1 | XP_017898043.1 | 5 - 6 | 1170 | 72 | ||
| XM_018042556.1 | XP_017898045.1 | 5 - 6 | 1170 | 72 | ||
| XM_018042557.1 | XP_017898046.1 | 5 - 6 | 1170 | 72 | ||
| XM_018042558.1 | XP_017898047.1 | 5 - 6 | 1167 | 72 | ||
| GenBank mRNA | JO630634.2 | - | 5 - 6 | 1047 | 72 | |
| JO630635.2 | - | 5 - 6 | 1044 | 72 | ||
More Information
Set Membership: |
|

NRG3
Genomic Map