Assay Details
| Gene Symbol: MRPL47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675281.3 | XP_005675338.1 | 6 - 7 | 693 | 62 | |
| XM_005675284.3 | XP_005675341.1 | 6 - 7 | 789 | 62 | ||
| GenBank mRNA | JT706133.1 | - | 7 - 8 | 904 | 62 | |
More Information
Set Membership: |
|

MRPL47
Genomic Map