Assay Details
| Gene Symbol: KBTBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689336.3 | XP_005689393.1 | 2 - 3 | 331 | 79 | |
| XM_013969383.2 | XP_013824837.1 | 2 - 3 | 288 | 79 | ||
| XM_018059904.1 | XP_017915393.1 | 2 - 3 | 345 | 79 | ||
| GenBank mRNA | JO607108.2 | - | 2 - 3 | 301 | 79 | |
More Information
Set Membership: |
|

KBTBD3
Genomic Map