Assay Details
| Gene Symbol: SYT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059199.1 | XP_017914688.1 | 5 - 6 | 1060 | 72 | |
| GenBank mRNA | JO646153.2 | - | 2 - 3 | 2994 | 72 | |
More Information
Set Membership: |
|

SYT13
Genomic Map