Assay Details
| Gene Symbol: AURKAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690784.3 | XP_005690841.1 | 3 - 4 | 699 | 65 | |
| XM_005690785.2 | XP_005690842.1 | 3 - 4 | 635 | 65 | ||
| GenBank mRNA | JO353835.2 | - | 3 - 4 | 616 | 65 | |
More Information
Set Membership: |
|

AURKAIP1
Genomic Map