Assay Details
| Gene Symbol: CENPQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696396.3 | XP_005696453.1 | 8 - 9 | 821 | 83 | |
| GenBank mRNA | JO443785.2 | - | 1 - 2 | 155 | 83 | |
More Information
Set Membership: |
|

CENPQ
Genomic Map