Assay Details
| Gene Symbol: C25H16orf72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040433.1 | XP_017895922.1 | 2 - 3 | 1002 | 85 | |
| GenBank mRNA | JO427355.1 | - | 1 - 2 | 1999 | 85 | |
More Information
Set Membership: |
|

C25H16orf72
Genomic Map