Assay Details
| Gene Symbol: TMEM72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699368.3 | XP_005699425.2 | 3 - 4 | 481 | 69 | |
| GenBank mRNA | JO429766.2 | - | 3 - 4 | 381 | 69 | |
More Information
Set Membership: |
|

TMEM72
Genomic Map