Assay Details
Gene Symbol: LOC102186671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045147.1 | XP_017900636.1 | 2258 | 67 | ||
GenBank mRNA | JO587095.2 | - | 1960 | 67 |
Gene Symbol: PHF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JT705290.1 | - | 3639 | 67 |
