Assay Details
Gene Symbol: LOC102186671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045147.1 | XP_017900636.1 | 3060 | 96 | ||
GenBank mRNA | JO587095.2 | - | 2762 | 96 |
Gene Symbol: PHF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043675.1 | XP_017899164.1 | 2 - 3 | 237 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
