Assay Details
| Gene Symbol: LOC102186671 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045147.1 | XP_017900636.1 | 3510 | 75 | ||
| GenBank mRNA | JO587095.2 | - | 3212 | 75 | ||
| Gene Symbol: PHF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043675.1 | XP_017899164.1 | 4 - 5 | 687 | 75 | |
| GenBank mRNA | JT705290.1 | - | 2374 | 75 | ||
More Information
Set Membership: |
|

LOC102186671
Genomic Map