Assay Details
| Gene Symbol: FMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013963528.2 | XP_013818982.1 | 1 - 2 | 281 | 62 | |
| GenBank mRNA | JO394118.2 | - | 1 - 2 | 340 | 62 | |
| JO598873.2 | - | 1 - 2 | 260 | 62 | ||
More Information
Set Membership: |
|

FMC1
Genomic Map