Assay Details
| Gene Symbol: ATN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048544.1 | XP_017904033.1 | 6 - 7 | 2805 | 80 | |
| XM_018048545.1 | XP_017904034.1 | 6 - 7 | 4659 | 80 | ||
| XM_018048546.1 | XP_017904035.1 | 6 - 7 | 2679 | 80 | ||
| XM_018048547.1 | XP_017904036.1 | 6 - 7 | 2802 | 80 | ||
| GenBank mRNA | KA330724.1 | - | 2 - 3 | 1823 | 80 | |
More Information
Set Membership: |
|

ATN1
Genomic Map