Assay Details
| Gene Symbol: FILIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052873.1 | XP_017908362.1 | 2 - 3 | 798 | 72 | |
| XM_018052874.1 | XP_017908363.1 | 2 - 3 | 346 | 72 | ||
| XM_018052875.1 | XP_017908364.1 | 2 - 3 | 798 | 72 | ||
More Information
Set Membership: |
|

FILIP1
Genomic Map