Assay Details
| Gene Symbol: CDK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013972144.2 | XP_013827598.1 | 5 - 6 | 705 | 69 | |
| XM_013972145.2 | XP_013827599.1 | 4 - 5 | 638 | 69 | ||
| XM_018063772.1 | XP_017919261.1 | 5 - 6 | 702 | 69 | ||
| XM_018063773.1 | XP_017919262.1 | 5 - 6 | 690 | 69 | ||
| GenBank mRNA | JO504710.2 | - | 1 - 2 | 152 | 69 | |
More Information
Set Membership: |
|

CDK3
Genomic Map