Assay Details
| Gene Symbol: RAI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064786.1 | XP_017920275.1 | 3 - 4 | 5750 | 79 | |
| XM_018064787.1 | XP_017920276.1 | 2 - 3 | 5680 | 79 | ||
| XM_018064788.1 | XP_017920277.1 | 2 - 3 | 5682 | 79 | ||
| XM_018064789.1 | XP_017920278.1 | 2 - 3 | 5590 | 79 | ||
More Information
Set Membership: |
|

RAI1
Genomic Map