Assay Details
| Gene Symbol: RAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065139.1 | XP_017920628.1 | 3 - 4 | 418 | 60 | |
| XM_018065140.1 | XP_017920629.1 | 3 - 4 | 375 | 60 | ||
| GenBank mRNA | JT676645.1 | - | 3 - 4 | 428 | 60 | |
| JT747507.1 | - | 4 - 5 | 826 | 60 | ||
More Information
Set Membership: |
|

RAMP2
Genomic Map